missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexylboronic Acid (contains varying amounts of Anhydride), TCI America™
$40.43 - $40.43
Chemical Identifiers
| CAS | 16343-08-1 |
|---|---|
| Molecular Formula | C6H15BO2 |
| MDL Number | MFCD01074641 |
| InChI Key | CXSYDLCMCLCOCA-UHFFFAOYSA-N |
| PubChem CID | 351064 |
| IUPAC Name | hexylboronic acid |
Chemical Identifiers
| 16343-08-1 | |
| MFCD01074641 | |
| 351064 |
| C6H15BO2 | |
| CXSYDLCMCLCOCA-UHFFFAOYSA-N | |
| hexylboronic acid |
Specifications
| 16343-08-1 | |
| C6H15BO2 | |
| 1 g | |
| CXSYDLCMCLCOCA-UHFFFAOYSA-N | |
| hexylboronic acid | |
| 351064 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD01074641 | |
| n-hexylboronic acid, 1-hexaneboronic acid, boronic acid, b-hexyl, 1-hexylboronic acid, n-hexaneboronic acid, boronic acid, hexyl, b-hexyl-boronic acid, n-hexylboronicacid, 1-hexylboronic acid c6-b oh 2 | |
| B(CCCCCC)(O)O | |
| 129.994 | |
| 129.99 | |
| Hexylboronic Acid (contains varying amounts of Anhydride) |
Safety and Handling
TSCA : No