missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexylbenzene 98.0+%, TCI America™
Supplier: TCI America H0136250ML
Specifications
| Hexylbenzene | |
| -61°C | |
| 226°C | |
| MFCD00009526 | |
| 3082 | |
| LTEQMZWBSYACLV-UHFFFAOYSA-N | |
| hexylbenzene | |
| 14109 | |
| ≥98.0% (GC) |
| 1077-16-3 | |
| Colorless | |
| C12H18 | |
| 250 mL | |
| 1-phenylhexane, n-hexylbenzene, benzene, hexyl, hexane, 1-phenyl, unii-8e3d1a2bzp, 8e3d1a2bzp, phenylhexane, hexyl-benzen, 1-hexylbenzene, 4-hexylbenzene | |
| CCCCCCC1=CC=CC=C1 | |
| 162.28 | |
| 162.28 | |
| Liquid |
Chemical Identifiers
| 1077-16-3 | |
| 162.28 | |
| LTEQMZWBSYACLV-UHFFFAOYSA-N | |
| 14109 | |
| CCCCCCC1=CC=CC=C1 |
| C12H18 | |
| MFCD00009526 | |
| 1-phenylhexane, n-hexylbenzene, benzene, hexyl, hexane, 1-phenyl, unii-8e3d1a2bzp, 8e3d1a2bzp, phenylhexane, hexyl-benzen, 1-hexylbenzene, 4-hexylbenzene | |
| hexylbenzene |
Safety and Handling
EINECSNumber : (3)-0021
TSCA : Yes