missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexyl Propionate 98.0+%, TCI America™
$71.23 - $202.15
Chemical Identifiers
| CAS | 2445-76-3 |
|---|---|
| Molecular Formula | C9H18O2 |
| Molecular Weight (g/mol) | 158.241 |
| MDL Number | MFCD00085197 |
| InChI Key | GOKKOFHHJFGZHW-UHFFFAOYSA-N |
| Synonym | Propionic Acid Hexyl Ester |
| PubChem CID | 88454 |
| ChEBI | CHEBI:87549 |
| IUPAC Name | hexyl propanoate |
| SMILES | CCCCCCOC(=O)CC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
H133025ML
|
TCI America
H133025ML |
25 mL |
Each for $71.23
|
|
|||||
|
H1330500ML
|
TCI America
H1330500ML |
500 mL |
Each for $202.15
|
|
|||||
Chemical Identifiers
| 2445-76-3 | |
| 158.241 | |
| GOKKOFHHJFGZHW-UHFFFAOYSA-N | |
| 88454 | |
| hexyl propanoate |
| C9H18O2 | |
| MFCD00085197 | |
| Propionic Acid Hexyl Ester | |
| CHEBI:87549 | |
| CCCCCCOC(=O)CC |
Specifications
| 2445-76-3 | |
| 186°C | |
| MFCD00085197 | |
| Propionic Acid Hexyl Ester | |
| CCCCCCOC(=O)CC | |
| 158.241 | |
| CHEBI:87549 | |
| ≥98.0% (GC) | |
| Hexyl Propionate |
| Colorless | |
| C9H18O2 | |
| 25 mL | |
| GOKKOFHHJFGZHW-UHFFFAOYSA-N | |
| hexyl propanoate | |
| 88454 | |
| 158.24 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0774
RTECSNumber : UA2462200
TSCA : Yes