missing translation for 'onlineSavingsMsg'
Learn More

Hexyl Methanesulfonate 96.0+%, TCI America™

Catalog No. H12825G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12825G 5 g
H128225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H12825G Supplier TCI America Supplier No. H12825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16156-50-6
Molecular Formula C7H16O3S
Molecular Weight (g/mol) 180.26
MDL Number MFCD00674707
InChI Key URIRDRHUUFRHAS-UHFFFAOYSA-N
Synonym Methanesulfonic Acid Hexyl Ester
PubChem CID 12517373
IUPAC Name hexyl methanesulfonate
SMILES CCCCCCOS(C)(=O)=O

Specifications

Color Yellow
Boiling Point 90°C
Quantity 5 g
Formula Weight 180.26
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexyl Methanesulfonate