missing translation for 'onlineSavingsMsg'
Learn More

Hexyl Methacrylate (stabilized with MEHQ) 98.0+%, TCI America™

Catalog No. M050325ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M050325ML 25 mL
M0503500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M050325ML Supplier TCI America Supplier No. M050325ML
Only null left

Chemical Identifiers

CAS 142-09-6
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.25
MDL Number MFCD00015283
InChI Key LNCPIMCVTKXXOY-UHFFFAOYSA-N
Synonym hexyl methacrylate, n-hexyl methacrylate, hexyl 2-methyl-2-propenoate, methacrylic acid, hexyl ester, 2-propenoic acid, 2-methyl-, hexyl ester, unii-538b2q3hsv, ccris 4824, hexyl 2-methylacrylate, methacrylic acid hexyl ester, dsstox_cid_5406
PubChem CID 8872
IUPAC Name hexyl 2-methylprop-2-enoate
SMILES CCCCCCOC(=O)C(C)=C

Specifications

Color Colorless
Boiling Point 88°C
Quantity 25 mL
Formula Weight 170.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexyl Methacrylate (stabilized with MEHQ)
EINECSNumber (2)-1039
TSCA Yes