missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexyl Isocyanate 98.0+%, TCI America™
Supplier: TCI America I049225ML
Specifications
| Hexyl Isocyanate | |
| Yellow | |
| C7H13NO | |
| 25 mL | |
| hexyl isocyanate, hexane, 1-isocyanato, 1-hexyl isocyanate, hexylmonoisocyanate, hexyl monoisocyanate, hexylisocyanate, n-hexyl isocyanate, hexanisocyanate, isocyanatohexane, isocyanato-hexane | |
| CCCCCCN=C=O | |
| 127.19 | |
| CHEBI:59058 | |
| ≥98.0% (GC) |
| 2525-62-4 | |
| 164°C | |
| MFCD00037047 | |
| 3080 | |
| ANJPRQPHZGHVQB-UHFFFAOYSA-N | |
| 1-isocyanatohexane | |
| 75659 | |
| 127.19 | |
| Liquid |
Chemical Identifiers
| 2525-62-4 | |
| 127.19 | |
| ANJPRQPHZGHVQB-UHFFFAOYSA-N | |
| 75659 | |
| 1-isocyanatohexane |
| C7H13NO | |
| MFCD00037047 | |
| hexyl isocyanate, hexane, 1-isocyanato, 1-hexyl isocyanate, hexylmonoisocyanate, hexyl monoisocyanate, hexylisocyanate, n-hexyl isocyanate, hexanisocyanate, isocyanatohexane, isocyanato-hexane | |
| CHEBI:59058 | |
| CCCCCCN=C=O |
Safety and Handling
TSCA : Yes
Recommended Storage : Refrigerator