missing translation for 'onlineSavingsMsg'
Learn More

Hexyl Isocyanate 98.0+%, TCI America™

Catalog No. I04925ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04925ML 5 mL
I049225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I04925ML Supplier TCI America Supplier No. I04925ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2525-62-4
Molecular Formula C7H13NO
Molecular Weight (g/mol) 127.19
MDL Number MFCD00037047
InChI Key ANJPRQPHZGHVQB-UHFFFAOYSA-N
Synonym hexyl isocyanate, hexane, 1-isocyanato, 1-hexyl isocyanate, hexylmonoisocyanate, hexyl monoisocyanate, hexylisocyanate, n-hexyl isocyanate, hexanisocyanate, isocyanatohexane, isocyanato-hexane
PubChem CID 75659
ChEBI CHEBI:59058
IUPAC Name 1-isocyanatohexane
SMILES CCCCCCN=C=O

Specifications

Color Yellow
Boiling Point 164°C
Quantity 5 mL
UN Number 3080
Formula Weight 127.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexyl Isocyanate
TSCA Yes
Recommended Storage Refrigerator