Learn More
Hexyl isocyanate, 99%
CAS: 2525-62-4 | C7H13NO | 127.19 g/mol
Supplier: Thermo Scientific Chemicals 411660250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Hexyl isocyanate | |
| 2525-62-4 | |
| 162°C to 164°C | |
| Authentic | |
| Glass bottle | |
| 1.4190 to 1.421 | |
| MFCD00037047 | |
| 04, IV, 718 | |
| Solubility in water: decomposes | |
| CCCCCCN=C=O | |
| 127.19 | |
| CHEBI:59058 | |
| 99% |
| 99% | |
| Colorless to Yellow | |
| 59°C | |
| 98.5% min. (GC) | |
| C7H13NO | |
| CH3(CH2)5NCO | |
| 25 g | |
| hexyl isocyanate, hexane, 1-isocyanato, 1-hexyl isocyanate, hexylmonoisocyanate, hexyl monoisocyanate, hexylisocyanate, n-hexyl isocyanate, hexanisocyanate, isocyanatohexane, isocyanato-hexane | |
| ANJPRQPHZGHVQB-UHFFFAOYSA-N | |
| 1-isocyanatohexane | |
| 75659 | |
| 127.19 | |
| Liquid |
Chemical Identifiers
| 2525-62-4 | |
| 127.19 | |
| ANJPRQPHZGHVQB-UHFFFAOYSA-N | |
| 75659 | |
| 1-isocyanatohexane |
| C7H13NO | |
| MFCD00037047 | |
| hexyl isocyanate, hexane, 1-isocyanato, 1-hexyl isocyanate, hexylmonoisocyanate, hexyl monoisocyanate, hexylisocyanate, n-hexyl isocyanate, hexanisocyanate, isocyanatohexane, isocyanato-hexane | |
| CHEBI:59058 | |
| CCCCCCN=C=O |
Safety and Handling
GHS H Statement
Harmful if swallowed. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Harmful if inhaled. May cause respiratory irritation. Causes serious eye damage. Harmful in contact with skin. Causes skin irritation. Flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Avoid breathing dust/fume/gas/mist/vapors/spray. IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing. If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
GHS Signal Word: Danger
EINECSNumber : 219-763-8
TSCA : TSCA
Recommended Storage : Product may develop some turbidity or precipitate
RUO – Research Use Only