missing translation for 'onlineSavingsMsg'
Learn More

Hexyl isocyanate, 99%

Catalog No. AC411661000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC411661000 100 g Glass bottle
AC411660050 5 g Glass bottle
AC411660250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC411661000 Supplier Thermo Scientific Chemicals Supplier No. 411661000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2525-62-4 | C7H13NO | 127.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2525-62-4
Molecular Formula C7H13NO
Molecular Weight (g/mol) 127.19
MDL Number MFCD00037047
InChI Key ANJPRQPHZGHVQB-UHFFFAOYSA-N
Synonym hexyl isocyanate, hexane, 1-isocyanato, 1-hexyl isocyanate, hexylmonoisocyanate, hexyl monoisocyanate, hexylisocyanate, n-hexyl isocyanate, hexanisocyanate, isocyanatohexane, isocyanato-hexane
PubChem CID 75659
ChEBI CHEBI:59058
IUPAC Name 1-isocyanatohexane
SMILES CCCCCCN=C=O

Specifications

Color Colorless to Yellow
Boiling Point 162°C to 164°C
Flash Point 59°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4190 to 1.421
Linear Formula CH3(CH2)5NCO
Quantity 100 g
Beilstein 04, IV, 718
Solubility Information Solubility in water: decomposes
Formula Weight 127.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Hexyl isocyanate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Harmful if inhaled. May cause respiratory irritation. Causes serious eye damage. Harmful in contact with skin. Causes skin irritation. Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Avoid breathing dust/fume/gas/mist/vapors/spray. IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing. If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
EINECSNumber 219-763-8
TSCA TSCA
Recommended Storage Product may develop some turbidity or precipitate

RUO – Research Use Only