missing translation for 'onlineSavingsMsg'
Learn More

Hexyl Hexanoate 98.0+%, TCI America™

Catalog No. C151425ML
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
500 mL
Catalog No. Quantity
C151425ML 25 mL
C1514500ML 500 mL
2 options

Catalog No. C151425ML

Supplier: TCI America C151425ML

Only null left

Chemical Identifiers

CAS 6378-65-0
Molecular Formula C12H24O2
Molecular Weight (g/mol) 200.32
MDL Number MFCD00053808
InChI Key NCDCLPBOMHPFCV-UHFFFAOYSA-N
Synonym Hexanoic Acid Hexyl Ester
PubChem CID 22873
IUPAC Name hexyl hexanoate
SMILES CCCCCCOC(=O)CCCCC

Specifications

Color Colorless
Boiling Point 244°C
Quantity 25 mL
Formula Weight 200.32
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexyl Hexanoate
EINECSNumber (2)-0760
RTECSNumber MO8385000
TSCA Yes