missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexyl Chloroformate 98.0+%, TCI America™
$155.93 - $506.33
Chemical Identifiers
| CAS | 6092-54-2 |
|---|---|
| Molecular Formula | C7H13ClO2 |
| Molecular Weight (g/mol) | 164.63 |
| MDL Number | MFCD00000651 |
| InChI Key | KIWBRXCOTCXSSZ-UHFFFAOYSA-N |
| Synonym | hexyl chloroformate, carbonochloridic acid, hexyl ester, n-hexyl chloroformate, chloroformic acid n-hexyl ester, hexyl chlorocarbonate, unii-mu6004c2z3, chloro hexyloxy methanone, chloroformic acid hexyl ester, hexylchloroformate, hexyl=chloroformate |
| PubChem CID | 22466 |
| IUPAC Name | hexyl carbonochloridate |
| SMILES | CCCCCCOC(Cl)=O |
Chemical Identifiers
| 6092-54-2 | |
| 164.63 | |
| KIWBRXCOTCXSSZ-UHFFFAOYSA-N | |
| 22466 | |
| CCCCCCOC(Cl)=O |
| C7H13ClO2 | |
| MFCD00000651 | |
| hexyl chloroformate, carbonochloridic acid, hexyl ester, n-hexyl chloroformate, chloroformic acid n-hexyl ester, hexyl chlorocarbonate, unii-mu6004c2z3, chloro hexyloxy methanone, chloroformic acid hexyl ester, hexylchloroformate, hexyl=chloroformate | |
| hexyl carbonochloridate |
Specifications
| 6092-54-2 | |
| 78°C | |
| MFCD00000651 | |
| 3277 | |
| KIWBRXCOTCXSSZ-UHFFFAOYSA-N | |
| hexyl carbonochloridate | |
| 22466 | |
| ≥98.0% (GC,T) | |
| Hexyl Chloroformate |
| Colorless | |
| C7H13ClO2 | |
| 5 mL | |
| hexyl chloroformate, carbonochloridic acid, hexyl ester, n-hexyl chloroformate, chloroformic acid n-hexyl ester, hexyl chlorocarbonate, unii-mu6004c2z3, chloro hexyloxy methanone, chloroformic acid hexyl ester, hexylchloroformate, hexyl=chloroformate | |
| CCCCCCOC(Cl)=O | |
| 164.63 | |
| 164.63 | |
| Liquid |
Safety and Handling
TSCA : Yes