missing translation for 'onlineSavingsMsg'
Learn More

Hexyl Chloroformate 98.0+%, TCI America™

Catalog No. C07235ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C07235ML 5 mL
C072325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C07235ML Supplier TCI America Supplier No. C07235ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6092-54-2
Molecular Formula C7H13ClO2
Molecular Weight (g/mol) 164.63
MDL Number MFCD00000651
InChI Key KIWBRXCOTCXSSZ-UHFFFAOYSA-N
Synonym hexyl chloroformate, carbonochloridic acid, hexyl ester, n-hexyl chloroformate, chloroformic acid n-hexyl ester, hexyl chlorocarbonate, unii-mu6004c2z3, chloro hexyloxy methanone, chloroformic acid hexyl ester, hexylchloroformate, hexyl=chloroformate
PubChem CID 22466
IUPAC Name hexyl carbonochloridate
SMILES CCCCCCOC(Cl)=O

Specifications

Color Colorless
Boiling Point 78°C
Quantity 5 mL
UN Number 3277
Formula Weight 164.63
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Hexyl Chloroformate