Learn More
Hexyl chloroformate, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 427040250
| Quantity | 25g |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Coupling reagentChemical Identifiers
| C7H13ClO2 | |
| MFCD00000651 | |
| hexyl chloroformate, carbonochloridic acid, hexyl ester, n-hexyl chloroformate, chloroformic acid n-hexyl ester, hexyl chlorocarbonate, unii-mu6004c2z3, chloro hexyloxy methanone, chloroformic acid hexyl ester, hexylchloroformate, hexyl=chloroformate | |
| hexyl carbonochloridate |
Specifications
| Hexyl chloroformate | |
| 1.0070g/mL | |
| 62°C | |
| 97.5% min. (GC) | |
| C7H13ClO2 | |
| ClCO2(CH2)5CH3 | |
| MFCD00000651 | |
| hexyl chloroformate, carbonochloridic acid, hexyl ester, n-hexyl chloroformate, chloroformic acid n-hexyl ester, hexyl chlorocarbonate, unii-mu6004c2z3, chloro hexyloxy methanone, chloroformic acid hexyl ester, hexylchloroformate, hexyl=chloroformate | |
| CCCCCCOC(Cl)=O | |
| 164.63 | |
| 164.63 |
| 6092-54-2 | |
| 60°C to 61°C (7.0mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.4230 to 1.4250 (20°C, 589nm) | |
| 25g | |
| 1.007 | |
| KIWBRXCOTCXSSZ-UHFFFAOYSA-N | |
| hexyl carbonochloridate | |
| 22466 | |
| 98% |
Safety and Handling
GHS H Statement:
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Toxic if inhaled.
Contact with water liberates toxic gas.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 228-036-4