missing translation for 'onlineSavingsMsg'
Learn More

Hexyl Butyrate 98.0+%, TCI America™

Catalog No. B1891500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1891500ML 500 mL
B189125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1891500ML Supplier TCI America Supplier No. B1891500ML
Only null left

Chemical Identifiers

CAS 2639-63-6
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.27
MDL Number MFCD00048884
InChI Key XAPCMTMQBXLDBB-UHFFFAOYSA-N
Synonym Butyric Acid Hexyl Ester
PubChem CID 17525
ChEBI CHEBI:87559
IUPAC Name hexyl butanoate
SMILES CCCCCCOC(=O)CCC

Specifications

Color Colorless
Boiling Point 205°C
Quantity 500 mL
Formula Weight 172.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexyl Butyrate
EINECSNumber (2)-0779
RTECSNumber ET4203000
TSCA Yes