missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexyl Acetate 99.0+%, TCI America™
$31.68 - $102.52
Chemical Identifiers
| CAS | 142-92-7 |
|---|---|
| Molecular Formula | C8H16O2 |
| Molecular Weight (g/mol) | 144.214 |
| MDL Number | MFCD00009524 |
| InChI Key | AOGQPLXWSUTHQB-UHFFFAOYSA-N |
| Synonym | n-hexyl acetate, 1-hexyl acetate, hexyl ethanoate, acetic acid hexyl ester, n-hexyl ethanoate, acetic acid, hexyl ester, hexyl alcohol, acetate, l-hexyl acetate, fema no. 2565, natural |
| PubChem CID | 8908 |
| ChEBI | CHEBI:87510 |
| IUPAC Name | hexyl acetate |
| SMILES | CCCCCCOC(=O)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A003225ML
|
TCI America
A003225ML |
25 mL |
Each for $31.68
|
|
|||||
|
A0032500ML
|
TCI America
A0032500ML |
500 mL |
Each for $102.52
|
|
|||||
Chemical Identifiers
| 142-92-7 | |
| 144.214 | |
| AOGQPLXWSUTHQB-UHFFFAOYSA-N | |
| 8908 | |
| hexyl acetate |
| C8H16O2 | |
| MFCD00009524 | |
| n-hexyl acetate, 1-hexyl acetate, hexyl ethanoate, acetic acid hexyl ester, n-hexyl ethanoate, acetic acid, hexyl ester, hexyl alcohol, acetate, l-hexyl acetate, fema no. 2565, natural | |
| CHEBI:87510 | |
| CCCCCCOC(=O)C |
Specifications
| 142-92-7 | |
| 169°C | |
| MFCD00009524 | |
| n-hexyl acetate, 1-hexyl acetate, hexyl ethanoate, acetic acid hexyl ester, n-hexyl ethanoate, acetic acid, hexyl ester, hexyl alcohol, acetate, l-hexyl acetate, fema no. 2565, natural | |
| CCCCCCOC(=O)C | |
| 144.214 | |
| CHEBI:87510 | |
| ≥99.0% (GC) | |
| Hexyl Acetate |
| Colorless | |
| C8H16O2 | |
| 25 mL | |
| AOGQPLXWSUTHQB-UHFFFAOYSA-N | |
| hexyl acetate | |
| 8908 | |
| 144.21 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0734&(2)-0735
RTECSNumber : AI0875000
TSCA : Yes