missing translation for 'onlineSavingsMsg'
Learn More

Hexyl Acetate 99.0+%, TCI America™

Catalog No. A003225ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A003225ML 25 mL
A0032500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A003225ML Supplier TCI America Supplier No. A003225ML

Chemical Identifiers

CAS 142-92-7
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
MDL Number MFCD00009524
InChI Key AOGQPLXWSUTHQB-UHFFFAOYSA-N
Synonym n-hexyl acetate, 1-hexyl acetate, hexyl ethanoate, acetic acid hexyl ester, n-hexyl ethanoate, acetic acid, hexyl ester, hexyl alcohol, acetate, l-hexyl acetate, fema no. 2565, natural
PubChem CID 8908
ChEBI CHEBI:87510
IUPAC Name hexyl acetate
SMILES CCCCCCOC(=O)C

Specifications

Color Colorless
Boiling Point 169°C
Quantity 25 mL
Formula Weight 144.21
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexyl Acetate
EINECSNumber (2)-0734&(2)-0735
RTECSNumber AI0875000
TSCA Yes