missing translation for 'onlineSavingsMsg'
Learn More

HexArmor™ PointGuard™ Ultra 6044 Gloves

Catalog No. 19161136A
Encompass
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19161136A X-Small
19161136B Small
19161136C Medium
19161136D Large
19161136E X-Large
19161136F 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19161136A Supplier HexArmor™ Supplier No. 6044XS
Only null left

SuperFabric™ brand material offers highest level of cut-resistance on enhanced areas. Recommended for use as an under-glove solution with appropriate top-glove combination

  • High needlestick resistance provided by layering SuperFabric™ brand material
  • Tested using actual 25 gauge needles
  • Lightweight spandex shell with elastic wrist
  • Launderable

Specifications

Certifications/Compliance ANSI/ISEA 105 standard
Color Black
Cut Resistance Rating ANSI A9
Material SuperFabric
Product Type Cut-resistant Gloves
Size X-Small
EN388 Standard EN 388:4X33F
For Use With (Application) waste/recycle sorting, hospital waste handling, laundry and linen, wastewater/sewage treatment, municipality work
Glove Cuff Style Elasticated
Model 6044-XS
Series PointGuard™ Ultra 6044
Show More Show Less