Learn More
HexArmor™ Hercules™ 400R6E Gloves DFS Item
Heavy duty, gauntlet style gloves protect hands from cuts, punctures and scratches in high-risk situations.
$614.69 - $646.95
Specifications
Color Gray
Cut Resistance Rating ANSI A9
Material SuperFabric
Product Type Cut-resistant Gloves
EN388 Standard EN 388:4X34X
View More Specs
Products 3
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-130-3731B DFS Item
View Documents
HexArmor™
400R6EM
Medium
Pair for $646.95
Only null left
19-130-3731C DFS Item
View Documents
HexArmor™
400R6EL
Large
Pair for $646.95
Only null left
19-130-3731D DFS Item
View Documents
HexArmor™
400R6EXL
X-Large
Pair for $614.69
Only null left
Description
Description
Suitable for use in a wide range of industrial and animal handling applications, including waste handling/recycle processing, meat packing, lab work, and hospital waste handling and laundering
  • Gauntlet cuff provides full protection for entire hand and forearm
  • Cut- and puncture-resistant1 palm and back of hand
  • Special gripping substance on palm enhances abrasion resistance and provides better grip
  • Double layered in high-risk areas
  • Fully lined interior
  • High-temp washable
Specifications
Specifications
Gray
SuperFabric
EN 388:4X34X
Gauntlet
Hercules™ 400R6E
ANSI A9
Cut-resistant Gloves
Industrial and animal handling applications, including waste handling/recycle processing, meat packing, lab work and hospital waste handling and laundering
Silicone
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'