Learn More
HexArmor™ Hercules™ 400R6E Gloves
Catalog No. 191303731B
Encompass
Change view
Click to view available options
Size:
Medium
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
191303731B Medium
191303731C Large
191303731D X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 191303731B Supplier HexArmor™ Supplier No. 400R6EM
Heavy duty, gauntlet style gloves protect hands from cuts, punctures and scratches in high-risk situations.
Suitable for use in a wide range of industrial and animal handling applications, including waste handling/recycle processing, meat packing, lab work, and hospital waste handling and laundering
  • Gauntlet cuff provides full protection for entire hand and forearm
  • Cut- and puncture-resistant1 palm and back of hand
  • Special gripping substance on palm enhances abrasion resistance and provides better grip
  • Double layered in high-risk areas
  • Fully lined interior
  • High-temp washable
Specifications
Color Gray
Cut Resistance Rating ANSI A9
Material SuperFabric
Product Type Cut-resistant Gloves
Size Medium
EN388 Standard EN 388:4X34X
For Use With (Application) Industrial and animal handling applications, including waste handling/recycle processing, meat packing, lab work and hospital waste handling and laundering
Glove Cuff Style Gauntlet
Grip Design Silicone
Model 400R6E-M
Series Hercules™ 400R6E
Show More Show Less