missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexaphenyldisilane 97.0+%, TCI America™
$121.16 - $479.06
Chemical Identifiers
| CAS | 1450-23-3 |
|---|---|
| Molecular Formula | C36H30Si2 |
| Molecular Weight (g/mol) | 518.81 |
| MDL Number | MFCD00039558 |
| InChI Key | ZMHATUZXFSOVSC-UHFFFAOYSA-N |
| PubChem CID | 74059 |
| IUPAC Name | hexaphenyldisilane |
| SMILES | C1=CC=C(C=C1)[Si](C1=CC=CC=C1)(C1=CC=CC=C1)[Si](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
Chemical Identifiers
| 1450-23-3 | |
| 518.81 | |
| ZMHATUZXFSOVSC-UHFFFAOYSA-N | |
| hexaphenyldisilane |
| C36H30Si2 | |
| MFCD00039558 | |
| 74059 | |
| C1=CC=C(C=C1)[Si](C1=CC=CC=C1)(C1=CC=CC=C1)[Si](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
Specifications
| 1450-23-3 | |
| C36H30Si2 | |
| 1 g | |
| ZMHATUZXFSOVSC-UHFFFAOYSA-N | |
| hexaphenyldisilane | |
| 74059 | |
| ≥97.0% (GC) | |
| Hexaphenyldisilane |
| White-Yellow | |
| MFCD00039558 | |
| hexaphenyldisilane, disilane, hexaphenyl, 1,1,1,2,2,2-hexaphenyldisilane, disilane, 1,1,1,2,2,2-hexaphenyl, disilane, hexaphenyl-,, acmc-1bxza, triphenyl triphenylsilyl silane, 1,1,1,2,2,2-hexaphenyldisilane #, disilane,1,1,1,2,2,2-hexaphenyl | |
| C1=CC=C(C=C1)[Si](C1=CC=CC=C1)(C1=CC=CC=C1)[Si](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 518.81 | |
| 518.81 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes