missing translation for 'onlineSavingsMsg'
Learn More

Hexaphenylbenzene, TCI America™

Catalog No. h14121g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
Catalog No. Quantity
H14121G 1 g
1 options

Catalog No. H14121G

Supplier: TCI America H14121G

Only null left

Chemical Identifiers

CAS 992-04-1
Molecular Formula C42H30
Molecular Weight (g/mol) 534.702
MDL Number MFCD00003057
InChI Key QBHWPVJPWQGYDS-UHFFFAOYSA-N
Synonym hexaphenylbenzene, benzene, hexaphenyl, 3',4',5',6'-tetraphenyl-o-terphenyl, acmc-20akri, m-terphenyl, 2',4',5',6'-tetraphenyl, ghl.pd_mitscher_leg0.890, hexaphenylbenzene 1g, 1,1':2',1-terphenyl, 3',4',5',6'-tetraphenyl, 1,2,3,4,5,6-hexaphenylbenzene
PubChem CID 70432
IUPAC Name 1,2,3,4,5,6-hexakis-phenylbenzene
SMILES C1=CC=C(C=C1)C2=C(C(=C(C(=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5)C6=CC=CC=C6)C7=CC=CC=C7

Specifications

Melting Point 455°C
Color White-Yellow
Quantity 1 g
Formula Weight 534.70
Physical Form Crystalline Powder
Chemical Name or Material Hexaphenylbenzene