missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexaphenylbenzene, 98+%
Description
Hexaphenylbenzene was used to prepare the fluorescent nanorods used for the detection of trinitrotoulene (TNT).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Hexaphenylbenzene was used to prepare the fluorescent nanorods used for the detection of trinitrotoulene (TNT).
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Hexaphenylbenzene was used to prepare the fluorescent nanorods used for the detection of trinitrotoulene (TNT).
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | >300°C |
| Quantity | 1 g |
| Beilstein | 1895903 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 534.7 |
| Percent Purity | ≥98% |
| Chemical Name or Material | Hexaphenylbenzene |
Safety and Handling
| EINECSNumber | 213-591-7 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only