missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexanophenone 98.0+%, TCI America™
$130.92 - $1029.97
Chemical Identifiers
| CAS | 942-92-7 |
|---|---|
| Molecular Formula | C12H16O |
| Molecular Weight (g/mol) | 176.26 |
| MDL Number | MFCD00009512 |
| InChI Key | MAHPVQDVMLWUAG-UHFFFAOYSA-N |
| Synonym | hexanophenone, caprophenone, 1-hexanone, 1-phenyl, 1-phenyl-1-hexanone, n-hexanophenone, amyl phenyl ketone, hexaphenone, pentyl phenyl ketone, 4-ethylbutyrophenone, n-amylphenyl ketone |
| PubChem CID | 70337 |
| IUPAC Name | 1-phenylhexan-1-one |
| SMILES | CCCCCC(=O)C1=CC=CC=C1 |
Chemical Identifiers
| 942-92-7 | |
| 176.26 | |
| MAHPVQDVMLWUAG-UHFFFAOYSA-N | |
| 70337 | |
| CCCCCC(=O)C1=CC=CC=C1 |
| C12H16O | |
| MFCD00009512 | |
| hexanophenone, caprophenone, 1-hexanone, 1-phenyl, 1-phenyl-1-hexanone, n-hexanophenone, amyl phenyl ketone, hexaphenone, pentyl phenyl ketone, 4-ethylbutyrophenone, n-amylphenyl ketone | |
| 1-phenylhexan-1-one |
Specifications
| 942-92-7 | |
| C12H16O | |
| 25 g | |
| MAHPVQDVMLWUAG-UHFFFAOYSA-N | |
| 1-phenylhexan-1-one | |
| 70337 | |
| ≥98.0% (GC) | |
| Hexanophenone |
| White-Yellow | |
| MFCD00009512 | |
| hexanophenone, caprophenone, 1-hexanone, 1-phenyl, 1-phenyl-1-hexanone, n-hexanophenone, amyl phenyl ketone, hexaphenone, pentyl phenyl ketone, 4-ethylbutyrophenone, n-amylphenyl ketone | |
| CCCCCC(=O)C1=CC=CC=C1 | |
| 176.26 | |
| 176.26 | |
| Crystalline Lumps |
Safety and Handling
TSCA : Yes