missing translation for 'onlineSavingsMsg'
Learn More

Hexanophenone 98.0+%, TCI America™

Catalog No. H0116500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0116500G 500 g
H011625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0116500G Supplier TCI America Supplier No. H0116500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 942-92-7
Molecular Formula C12H16O
Molecular Weight (g/mol) 176.26
MDL Number MFCD00009512
InChI Key MAHPVQDVMLWUAG-UHFFFAOYSA-N
Synonym hexanophenone, caprophenone, 1-hexanone, 1-phenyl, 1-phenyl-1-hexanone, n-hexanophenone, amyl phenyl ketone, hexaphenone, pentyl phenyl ketone, 4-ethylbutyrophenone, n-amylphenyl ketone
PubChem CID 70337
IUPAC Name 1-phenylhexan-1-one
SMILES CCCCCC(=O)C1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 500 g
Formula Weight 176.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Hexanophenone