missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexanoic Anhydride 96.0+%, TCI America™
$33.56 - $216.24
Chemical Identifiers
| CAS | 2051-49-2 |
|---|---|
| Molecular Formula | C12H22O3 |
| Molecular Weight (g/mol) | 214.305 |
| MDL Number | MFCD00009509 |
| InChI Key | PKHMTIRCAFTBDS-UHFFFAOYSA-N |
| Synonym | hexanoic anhydride, caproic anhydride, hexanoic acid, anhydride, n-hexanoic anhydride, hexanoic acid, 1,1'-anhydride, unii-gg70y8ltby, n-caproic anhydride, caproic acid anhydride, capronic acid anhydride, n-hexanoic acid anhydride |
| PubChem CID | 74918 |
| IUPAC Name | hexanoyl hexanoate |
| SMILES | CCCCCC(=O)OC(=O)CCCCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
C002125ML
|
TCI America
C002125ML |
25 mL |
Each for $33.56
|
|
|||||
|
C0021500ML
|
TCI America
C0021500ML |
500 mL |
Each for $216.24
|
|
|||||
Chemical Identifiers
| 2051-49-2 | |
| 214.305 | |
| PKHMTIRCAFTBDS-UHFFFAOYSA-N | |
| 74918 | |
| CCCCCC(=O)OC(=O)CCCCC |
| C12H22O3 | |
| MFCD00009509 | |
| hexanoic anhydride, caproic anhydride, hexanoic acid, anhydride, n-hexanoic anhydride, hexanoic acid, 1,1'-anhydride, unii-gg70y8ltby, n-caproic anhydride, caproic acid anhydride, capronic acid anhydride, n-hexanoic acid anhydride | |
| hexanoyl hexanoate |
Specifications
| 2051-49-2 | |
| 244°C | |
| MFCD00009509 | |
| 3265 | |
| PKHMTIRCAFTBDS-UHFFFAOYSA-N | |
| hexanoyl hexanoate | |
| 74918 | |
| ≥96.0% (GC) | |
| Hexanoic Anhydride |
| Yellow | |
| C12H22O3 | |
| 25 mL | |
| hexanoic anhydride, caproic anhydride, hexanoic acid, anhydride, n-hexanoic anhydride, hexanoic acid, 1,1'-anhydride, unii-gg70y8ltby, n-caproic anhydride, caproic acid anhydride, capronic acid anhydride, n-hexanoic acid anhydride | |
| CCCCCC(=O)OC(=O)CCCCC | |
| 214.305 | |
| 214.31 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0628
TSCA : Yes