Learn More
Hexanoic anhydride, 97%
CAS: 2051-49-2 | C12H22O3 | 214.3 g/mol
$41.03 - $130.08
Chemical Identifiers
| CAS | 2051-49-2 |
|---|---|
| Molecular Formula | C12H22O3 |
| Molecular Weight (g/mol) | 214.3 |
| MDL Number | MFCD00009509 |
| InChI Key | PKHMTIRCAFTBDS-UHFFFAOYSA-N |
| Synonym | hexanoic anhydride, caproic anhydride, hexanoic acid, anhydride, n-hexanoic anhydride, hexanoic acid, 1,1'-anhydride, unii-gg70y8ltby, n-caproic anhydride, caproic acid anhydride, capronic acid anhydride, n-hexanoic acid anhydride |
| PubChem CID | 74918 |
| IUPAC Name | hexanoyl hexanoate |
| SMILES | CCCCCC(=O)OC(=O)CCCCC |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC186750500
|
Thermo Scientific Chemicals
186750500 |
50 g | Glass bottle |
Each for $41.03
|
|
||||
|
AC186752500
|
Thermo Scientific Chemicals
186752500 |
250 g | Glass bottle |
Each for $130.08
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2051-49-2 | |
| 214.3 | |
| PKHMTIRCAFTBDS-UHFFFAOYSA-N | |
| 74918 | |
| CCCCCC(=O)OC(=O)CCCCC |
| C12H22O3 | |
| MFCD00009509 | |
| hexanoic anhydride, caproic anhydride, hexanoic acid, anhydride, n-hexanoic anhydride, hexanoic acid, 1,1'-anhydride, unii-gg70y8ltby, n-caproic anhydride, caproic acid anhydride, capronic acid anhydride, n-hexanoic acid anhydride | |
| hexanoyl hexanoate |
Specifications
| 2051-49-2 | |
| Colorless to Yellow | |
| 246°C to 248°C | |
| Authentic | |
| Glass bottle | |
| 1.4270 to 1.429 | |
| MFCD00009509 | |
| 02, 324 | |
| hexanoic anhydride, caproic anhydride, hexanoic acid, anhydride, n-hexanoic anhydride, hexanoic acid, 1,1'-anhydride, unii-gg70y8ltby, n-caproic anhydride, caproic acid anhydride, capronic acid anhydride, n-hexanoic acid anhydride | |
| PKHMTIRCAFTBDS-UHFFFAOYSA-N | |
| hexanoyl hexanoate | |
| 74918 | |
| 97% | |
| Hexanoic anhydride, 99% |
| -40°C | |
| 0.9260g/mL | |
| 130°C | |
| 97% | |
| C12H22O3 | |
| [CH3(CH2)4CO]2O | |
| 50 g | |
| 0.926 | |
| Solubility in water: 72g/L. Other solubilities: soluble in ethanol and ether | |
| CCCCCC(=O)OC(=O)CCCCC | |
| 214.3 | |
| 214.3 | |
| Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 218-121-4
TSCA : TSCA
RUO – Research Use Only