missing translation for 'onlineSavingsMsg'
Learn More

Hexanes, technical, mixture of isomers

CAS: 92112-69-1 | C6H14

$116.94 - $923.93

Specifications

Chemical Name or Material Hexanes
Melting Point -95.0°C
Name Note Mixture of isomers
CAS 92112-69-1,110-54-3
Density 0.6590g/mL
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC210590010
View Documents
Thermo Scientific Chemicals
210590010
1 L Glass Bottle
Each for $116.94
Only null left
 
AC210590025
View Documents
Thermo Scientific Chemicals
210590025
2.5 L Glass Bottle
Each for $179.26
Only null left
 
AC210590050
View Documents
Thermo Scientific Chemicals
210590050
5 L Plastic Drum
Each for $312.06
Only null left
 
AC210590250
View Documents
Thermo Scientific Chemicals
210590250
25 L Metal Drum
Each for $923.93
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Hexanes
Mixture of isomers
0.6590g/mL
−22°C
1.3748 to 1.3810
0.659
CH3(CH2)4CH3
0.31 mPa.s (20°C)
14, 4694
Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether and chloroform
Technical
-95.0°C
92112-69-1,110-54-3
69.0°C
Liquid
0.002% max.
C6H14
MFCD00009520
01, 142
Hex
86.18
Videos
SDS
Documents

Documents

RUO – Research Use Only