missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexanes, MilliporeSigma™
Ideal for HPLC
$126.39 - $317.32
Chemical Identifiers
| CAS | 110-54-3 |
|---|---|
| Molecular Formula | C6H14 |
| Molecular Weight (g/mol) | 86.18 |
| MDL Number | MFCD02179311 |
| InChI Key | VLKZOEOYAKHREP-UHFFFAOYSA-N |
| Synonym | n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes |
| PubChem CID | 8058 |
| ChEBI | CHEBI:29021 |
| IUPAC Name | hexane |
| SMILES | CCCCCC |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
MHX02906
|
Technidata
HX02906 |
1 L | Glass Bottle |
Each for $126.39
|
|
||||
|
MHX02901
|
Technidata
HX02901 |
4 L | Glass Bottle |
Each for $277.89
|
|
||||
|
MHX0290P1
|
Technidata
HX0290P1 |
4 L | Poly-coated Glass Bottle |
Each for $317.32
|
|
||||
Chemical Identifiers
| 110-54-3 | |
| 86.18 | |
| VLKZOEOYAKHREP-UHFFFAOYSA-N | |
| 8058 | |
| hexane |
| C6H14 | |
| MFCD02179311 | |
| n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
| CHEBI:29021 | |
| CCCCCC |
Specifications
| -139.4°C | |
| 0.66g/cm3 at 20°C | |
| 1 L | |
| 224.85°C (436.7°F) | |
| Glass Bottle | |
| C6H14 | |
| MFCD02179311 | |
| Partially soluble in water | |
| CCCCCC | |
| 86.18 | |
| CHEBI:29021 | |
| 98.50% | |
| Hexanes |
| 110-54-3 | |
| 69°C | |
| -23.15°C (-9.7°F), Closed cup | |
| Charactristic | |
| Liquid | |
| CH3(CH2)4CH3 with 2- and 3-methylpentanes and methylcyclopentane | |
| n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
| VLKZOEOYAKHREP-UHFFFAOYSA-N | |
| hexane | |
| 8058 | |
| 86.18 | |
| HPLC |
Safety and Handling
DOTInformation : DOT Class: 3, Packaging Group: II
Recommended Storage : Room Temperature