missing translation for 'onlineSavingsMsg'
Learn More

Hexanes, MilliporeSigma™

Catalog No. MHX02906
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
Poly-coated Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MHX02906 1 L Glass Bottle
MHX02901 4 L Glass Bottle
MHX0290P1 4 L Poly-coated Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MHX02906 Supplier Technidata Supplier No. HX02906

Ideal for HPLC

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Melting Point -139.4°C
Density 0.66g/cm3 at 20°C
Quantity 1 L
Flash Point -23.15°C (-9.7°F), Closed cup
Ignition Point 224.85°C (436.7°F)
Odor Charactristic
Packaging Glass Bottle
Physical Form Liquid
Linear Formula CH3(CH2)4CH3 with 2- and 3-methylpentanes and methylcyclopentane
Boiling Point 69°C
Solubility Information Partially soluble in water
Formula Weight 86.18
Percent Purity 98.50%
Grade HPLC
Chemical Name or Material Hexanes
Show More Show Less
DOTInformation DOT Class: 3, Packaging Group: II
Recommended Storage Room Temperature