missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexanes, MilliporeSigma™
Specifications
Specifications
| Melting Point | -139.4°C |
| Density | 0.66g/cm3 at 20°C |
| Quantity | 4 L |
| Flash Point | -23.15°C (-9.7°F), Closed cup |
| Ignition Point | 224.85°C (436.7°F) |
| Odor | Charactristic |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Linear Formula | CH3(CH2)4CH3 with 2- and 3-methylpentanes and methylcyclopentane |
| Solubility Information | Partially soluble in water |
| Show More |
Safety and Handling
| DOTInformation | DOT Class: 3, Packaging Group: II |
| Recommended Storage | Room Temperature |