missing translation for 'onlineSavingsMsg'
Learn More

Hexanes, for analysis, mixture of isomers

Catalog No. p-4258732
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC390740010 1 L Glass Bottle
AC390740025 2.5 L Glass Bottle
AC390740100 10 L Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC390740010 Supplier Thermo Scientific Chemicals Supplier No. 390740010
Only null left

CAS: 92112-69-1 | C6H14

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Hexanes
Melting Point -95.0°C
CAS 92112-69-1, 110-54-3
Acidity 0.0002 meq/g max.
Density 0.6590g/mL
Boiling Point 69.0°C
Quantity 1 L
Flash Point −22°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3748 to 1.3810
Residue after Evaporation 0.0005% max.
Specific Gravity 0.659
Molecular Formula C6H14
Linear Formula CH3(CH2)4CH3
Viscosity 0.31 mPa.s (20°C)
Water 0.01% max.
Synonym Hex
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether and chloroform
Formula Weight 86.18
Grade Analytical
Show More Show Less

RUO – Research Use Only