missing translation for 'onlineSavingsMsg'
Learn More

Hexanes, ACS Reagent, Reagents

Catalog No. 01337941
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
15 gal.
200 L
Packaging:
Glass Bottle
Poly Bottle
Safety Coated Glass
Steel Drum
Steel Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01337941 15 gal. Steel Drum
01337940 200 L Steel Drum
01337939 20 L Steel Pail
01337938 4 L Poly Bottle
01337937 4 L Glass Bottle
01337936 4 L Safety Coated Glass
01337935 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 01337941 Supplier Labchem Inc Supplier No. C121390015E

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Hexanes, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
IUPAC Name hexane
SMILES CCCCCC

Specifications

Melting Point -95°C
Color Colorless
Quantity 15 gal.
Packaging Steel Drum
Physical Form Liquid
Specific Gravity 0.665
Boiling Point 68.7°C
Grade ACS
Chemical Name or Material Hexanes, ACS Reagent