missing translation for 'onlineSavingsMsg'
Learn More

Hexanes, ACS Reagent Grade, Ricca Chemical

Catalog No. p-7215402
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
4 L
Catalog No. Quantity
RSOH00104C 4 L
1 options

Catalog No. RSOH00104C

Supplier: Ricca Chemical Company RSOH00104C

Only null left

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
IUPAC Name hexane
SMILES CCCCCC

Specifications

CAS Max % 100
Boiling Point 69°C
Quantity 4 L
Packaging Amber Glass Bottle
Residue after Evaporation max. 0.001%
Percent Purity min. 98.5%
Grade ACS
Chemical Name or Material Hexanes
ShelfLife 60 months