missing translation for 'onlineSavingsMsg'
Learn More

Hexanes, ACS Reagent Grade, Ricca Chemical

Catalog No. RSOH00104C
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RSOH00104C 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RSOH00104C Supplier Ricca Chemical Company Supplier No. RSOH00104C
Only null left

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
IUPAC Name hexane
SMILES CCCCCC

Specifications

CAS Max % 100
Quantity 4 L
Packaging Amber Glass Bottle
Residue after Evaporation max. 0.001%
Boiling Point 69°C
Percent Purity min. 98.5%
Grade ACS
Chemical Name or Material Hexanes
ShelfLife 60 months