Learn More
Hexanes, 99%, for spectroscopy ACS, mixture of isomers
CAS: 92112-69-1 | C6H14
Supplier: Thermo Scientific Chemicals 411550025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopyEcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.
Specifications
| Hexanes | |
| Mixture of isomers, for spectroscopy | |
| 0.01 max. at 280nm, 0.01 max. at 400nm, 0.02 max. at 250nm, 0.04 max. at 240nm, 0.1 max. at 230nm, 0.2 max. at 220nm, 0.3 max. at 210nm, 1 max. at 195nm | |
| 69.0°C | |
| −22°C | |
| Liquid | |
| 0.001% max. | |
| 0.659 | |
| C6H14 | |
| MFCD00009520 | |
| 0.31 mPa.s (20°C) | |
| 14, 4694 | |
| Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether and chloroform | |
| 99% |
| -95.0°C | |
| 92112-69-1,110-54-3 | |
| 0.6590g/mL | |
| 2.5 L | |
| Glass Bottle | |
| 1.3748 to 1.3810 | |
| 99% | |
| 0.0003 meq/g max. | |
| CH3(CH2)4CH3 | |
| Vapor Pressure: 160mbar at 20°C | |
| 01, 142 | |
| Hex | |
| 86.18 | |
| ACS |
RUO – Research Use Only