missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexanes, 99%, pure, mixture of isomers
CAS: 92112-69-1 | C6H14
$144.40 - $144.40
Specifications
| Chemical Name or Material | Hexanes |
|---|---|
| Name Note | Mixture of Isomers |
| CAS | 92112-69-1,110-54-3 |
| Color | Colorless |
| Density | 0.6590g/mL |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC411540010
|
Thermo Scientific Chemicals
411540010 |
1 L | EcoSafPak™ Glass Bottle |
Each for $144.40
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Hexanes | |
| 92112-69-1,110-54-3 | |
| 0.6590g/mL | |
| Liquid | |
| CH3(CH2)4CH3 | |
| Vapor Pressure: 160mbar at 20°C | |
| 14, 4694 | |
| 86.18 |
| Mixture of Isomers | |
| Colorless | |
| Authentic | |
| 99% | |
| MFCD00009520 | |
| 01, 142 | |
| Hex | |
| Pure |
RUO – Research Use Only