missing translation for 'onlineSavingsMsg'
Learn More

Hexanes, 98.5% Pure (approx 64% n-Hexane, ACS Guaranteed Reagent), MilliporeSigma™

Catalog No. MHX02995
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MHX02995 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MHX02995 Supplier Technidata Supplier No. HX02995

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Color Colorless
Density 0.65
Quantity 4 L
Physical Form Liquid
Residue after Evaporation 0.001% max.
Linear Formula CH3(CH2)4CH3
Formula Weight 86.18
Grade ACS
Chemical Name or Material Hexanes, 98.5%, approx 64% n-Hexane
Recommended Storage Room Temperature