missing translation for 'onlineSavingsMsg'
Learn More

Hexanes, 98.5%, OmniSolv™, ACS, MilliporeSigma™

Catalog No. p-4382195
Change view
Click to view available options
Quantity:
500 mL
20 L
200 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MHX02996 500 mL
MHX02993 20 L
MHX029920 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MHX02996 Supplier Technidata Supplier No. HX02996
Only null left

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Color Colorless
Quantity 500 mL
Physical Form Liquid
Assay Percent Range 0.985
Formula Weight 86.18
Chemical Name or Material Hexanes, For ACS analysis