missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexanes, 98.5%, For GC Analysis, Non-UV, OmsiSolv, MilliporeSigma™
Ideal for gas chromatography
Supplier: Technidata HX02981
Specifications
| Hexanes | |
| 110-54-3 | |
| 0.66g/cm3 at 20°C | |
| 4 L | |
| 224.85°C (436.7°F) | |
| Glass Bottle | |
| 98.50% | |
| MFCD02179311 | |
| n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
| VLKZOEOYAKHREP-UHFFFAOYSA-N | |
| hexane | |
| 8058 | |
| 86.18 |
| -139.4°C | |
| Colorless | |
| 69°C | |
| -23.15°C (-9.7°F), Closed cup | |
| Characteristic | |
| Liquid | |
| C6H14 | |
| UN 1208 | |
| Partially soluble in water | |
| CCCCCC | |
| 86.18 | |
| CHEBI:29021 | |
| ACS |
Chemical Identifiers
| 110-54-3 | |
| 86.18 | |
| VLKZOEOYAKHREP-UHFFFAOYSA-N | |
| 8058 | |
| hexane |
| C6H14 | |
| MFCD02179311 | |
| n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
| CHEBI:29021 | |
| CCCCCC |
Safety and Handling
DOTInformation : DOT Class: 3, Packaging Group: II
Recommended Storage : Room Temperature