missing translation for 'onlineSavingsMsg'
Learn More

Hexanes, 99.9%, Extra Dry, mixture of isomers, AcroSeal™

Catalog No. p-216881
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC610371000 100 mL AcroSeal™ Glass Bottle
AC610370010 1 L AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC610371000 Supplier Thermo Scientific Chemicals Supplier No. 610371000
Only null left

CAS: 92112-69-1 | C6H14

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Hexanes
CAS 92112-69-1, 110-54-3
Density 0.6590g/mL
Quantity 100 mL
Identification Pass Test
Packaging AcroSeal™ Glass Bottle
Assay Percent Range ≤97%
Molecular Formula C6H14
Vapor Pressure Vapor Pressure: 160mbar at 20°C
Linear Formula CH3(CH2)4CH3
MDL Number MFCD00009520
Water 50ppm max.
Beilstein 01, 142
Merck Index 14, 4694
Synonym Hex
Formula Weight 86.18
Percent Purity ≤97%
Grade Extra Dry
Show More Show Less

RUO – Research Use Only