missing translation for 'onlineSavingsMsg'
Learn More

Hexanes, 95+%, for spectroscopy, mixture of isomers

CAS: 92112-69-1 | C6H14 | 86.18 g/mol

Supplier:  Thermo Scientific Chemicals 168570025

 View more versions of this product

Catalog No. AC168570025


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.

Specifications

Specifications

Hexanes
0.00015 meq/g max.
-95°C
69°C
(95% n-hexane approximately) (GC)
C6H14
CH3(CH2)4CH3
2.5 L
0.0005% max.
14, 4694
Solubility in water: insoluble. Other solubilities: soluble in alcohol,acetone,ether and chloroform
0.31 mPa.s (20°C)
160mbar at 20°C
Spectroscopy
92112-69-1,110-54-3
(1 cm cell vs water), 0.005 max. at 240nm, 0.005 max. at 250nm, 0.01 max. at 230nm, 0.025 max. at 220nm, 0.1 max. at 210nm, 0.3 max. at 200nm
0.6590g/mL
-22°C
Glass bottle
1.3748 to 1.381
MFCD00009520
01, 142
0.659
Hex
86.18
86.18
95+%
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only