missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexane 99.5+%, TCI America™
Specifications
Specifications
| Melting Point | -95°C |
| Color | Colorless |
| Quantity | 5 mL |
| Physical Form | Clear Liquid at 20°C |
| UN Number | 1208 |
| Boiling Point | 68°C |
| Formula Weight | 86.18 |
| Percent Purity | ≥99.5% (GC) |
| Chemical Name or Material | Hexane [Standard Material for GC] |
Safety and Handling
| EINECSNumber | (2)-0006 |
| RTECSNumber | MN9275000 |
| TSCA | Yes |