missing translation for 'onlineSavingsMsg'
Learn More

Hexane, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, >99.9% (>60% n-Hexane), Solstice

Catalog No. 6001075
Change view
Click to view available options
Quantity:
19 L
204 L
Packaging:
Metal Can
Metal Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6001075 204 L Metal Drum
6001074 19 L Metal Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001075 Supplier Solstice Supplier No. PP211204

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

CAS Min % >60.0000%
CAS Max % >60.00%, <40.0000%
Density 0.659 - 0.673 g/cm3
Quantity 204 L
Flash Point −26°C
Packaging Metal Drum
Water 0.01% max.
UN Number UN1208
Beilstein 1730733
Boiling Point 68.7°C
Solubility Information 1.4% in Water at 20°C
Formula Weight 86.18
Percent Purity >60%
Grade ACS
Assay by GC Analysis n-Hexane
Chemical Name or Material Hexane, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, >99.9% (>60% n-Hexane)
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Hexanes