missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexane, Laboratory Reagent, ≥95%, Honeywell™
Laboratory Reagent,≥95%
Supplier: Honeywell Chemicals 2087524X25L
Description
- ≥95% for general laboratory use.
Specifications
| Hexane | |
| 110-54-3 | |
| 0.659g/mL at 25°C | |
| 4 x 2.5 L | |
| −26°C (−14.8°F) | |
| Glass Bottle | |
| n20/D 1.375(lit.) | |
| CH3(CH2)4CH3 | |
| 132mmHg (20°C), 256mmHg (37.7°C), 5.2psi (37.7°C) | |
| 1730733 | |
| VLKZOEOYAKHREP-UHFFFAOYSA-N | |
| hexane | |
| 8058 | |
| 86.18g/mol | |
| Laboratory Reagent |
| -95°C | |
| Colorless | |
| 69°C | |
| 0.077 | |
| 453°F | |
| Liquid | |
| C6H14 | |
| MFCD02179311 | |
| UN1208 | |
| n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
| CCCCCC | |
| 86.18 | |
| CHEBI:29021 | |
| ≥95% |
Chemical Identifiers
| 110-54-3 | |
| 86.18 | |
| VLKZOEOYAKHREP-UHFFFAOYSA-N | |
| 8058 | |
| hexane |
| C6H14 | |
| MFCD02179311 | |
| n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
| CHEBI:29021 | |
| CCCCCC |
Safety and Handling
P210-P260-P280-P301 + P310-P370 + P378-P403 + P235
H225-H304-H315-H336-H361f-H373-H411
DOTInformation : Class 3 - PG 2 - Hexanes
EINECSNumber : 203-777-6
RTECSNumber : MN9275000