missing translation for 'onlineSavingsMsg'
Learn More

Hexane 96.0+%, TCI America™

Catalog No. H0405100ML
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0405100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H0405100ML Supplier TCI America Supplier No. H0405100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Melting Point -95°C
Color Colorless
Quantity 100 mL
Physical Form Clear Liquid at 20°C
UN Number 1208
Boiling Point 68°C
Formula Weight 86.18
Percent Purity ≥96.0% (GC)
Chemical Name or Material Hexane [for Liquid Paraffin Test]
EINECSNumber (2)-0006
RTECSNumber MN9275000
TSCA Yes