missing translation for 'onlineSavingsMsg'
Learn More

Hexane CHROMASOLV™, for HPLC, ≥97.0% (GC), Honeywell Riedel-de Haën™

Catalog No. p-7073691
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001046 1 L
6001047 2.5 L
6001050 4 x 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6001046 Supplier Honeywell Chemicals Supplier No. 348591L
Only null left

CHROMASOLV™, for HPLC,≥97.0% (GC)

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Melting Point -95°C
Density 0.659g/mL at 25°C
Boiling Point 69°C
Quantity 1 L
Refractive Index n20/D 1.375
Linear Formula CH3(CH2)4CH3
Vapor Pressure 256mmHg (37.7°C)
Beilstein 1730733
Formula Weight 86.18g/mol
Percent Purity ≥97.0% (GC)
Grade CHROMASOLV
Chemical Name or Material Hexane, for HPLC
Show More Show Less
Hazard Statement P201-P210-P273-P301 + P310-P308 + P313-P331
Precautionary Statement H225-H304-H315-H336-H361f-H373-H411
EINECSNumber 203-777-6
RTECSNumber MN9275000