missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexane CHROMASOLV™, for HPLC, ≥97.0% (GC), Solstice
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Specifications
Specifications
| Melting Point | -95°C |
| Density | 0.659g/mL at 25°C |
| Quantity | 2.5 L |
| Refractive Index | n20/D 1.375 |
| Vapor Pressure | 256mmHg (37.7°C) |
| Linear Formula | CH3(CH2)4CH3 |
| Beilstein | 1730733 |
| Boiling Point | 69°C |
| Formula Weight | 86.18g/mol |
| Percent Purity | ≥97.0% (GC) |
| Show More |
Safety and Handling
| Hazard Statement | P201-P210-P273-P301 + P310-P308 + P313-P331 |
| Precautionary Statement | H225-H304-H315-H336-H361f-H373-H411 |
| EINECSNumber | 203-777-6 |
| RTECSNumber | MN9275000 |