missing translation for 'onlineSavingsMsg'
Learn More

Hexane, CHROMASOLV™, for HPLC, ≥95%, Honeywell Riedel-de Haën™

CHROMASOLV™, for HPLC,≥95%

Supplier:  Honeywell Chemicals 2705046X1L

 View more versions of this product

Catalog No. 6001045


Only null left
Explore available promotions

Description

Description

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
Specifications

Specifications

Hexane
CHROMASOLV™, for HPLC, ≥95%
max. 0.02 at 240 nm, max. 0.01 at 250 nm, max. 0.01 at 400 nm, max. 1.0 at 195 nm, max. 0.30 at 210 nm, max. 0.10 at 220 nm, max. 0.05 at 230 nm
≤100.0000%
66°C to 69°C
−26°C
C6H14
MFCD02179311
1730733
VLKZOEOYAKHREP-UHFFFAOYSA-N
hexane
8058
86.18
HPLC
-95°C (literature)
110-54-3
≥90.0000%
0.659 g/mL at 25 °C(lit.)
6 x 1 L
Glass Bottle
CH3(CH2)4CH3
UN1208
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
CCCCCC
86.18
CHEBI:29021
>95%

Chemical Identifiers

110-54-3
86.18
VLKZOEOYAKHREP-UHFFFAOYSA-N
CHEBI:29021
CCCCCC
C6H14
MFCD02179311
8058
hexane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 1260 days from date of manufacture

DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Hexanes

EINECSNumber : 203-777-6

Recommended Storage : Room Temp

SDS
Documents

Documents

Promotions

Promotions