missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexane, CHROMASOLV™, for HPLC, ≥95%, Honeywell Riedel-de Haën™
CHROMASOLV™, for HPLC,≥95%
Supplier: Honeywell-Chromasolv 27050425L
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Specifications
| Hexane | |
| CHROMASOLV™, for HPLC, ≥95% | |
| max. 0.02 at 240 nm, max. 0.01 at 250 nm, max. 0.01 at 400 nm, max. 1.0 at 195 nm, max. 0.30 at 210 nm, max. 0.10 at 220 nm, max. 0.05 at 230 nm | |
| ≤100.0000% | |
| 66°C to 69°C | |
| −26°C | |
| C6H14 | |
| MFCD02179311 | |
| 1730733 | |
| VLKZOEOYAKHREP-UHFFFAOYSA-N | |
| hexane | |
| 8058 | |
| 86.18 | |
| HPLC |
| -95°C (literature) | |
| 110-54-3 | |
| ≥90.0000% | |
| 0.659Â g/mLÂ at 25Â °C(lit.) | |
| 2.5 L | |
| Glass Bottle | |
| CH3(CH2)4CH3 | |
| UN1208 | |
| n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
| CCCCCC | |
| 86.18 | |
| CHEBI:29021 | |
| >95% |
Chemical Identifiers
| 110-54-3 | |
| 86.18 | |
| VLKZOEOYAKHREP-UHFFFAOYSA-N | |
| CHEBI:29021 | |
| CCCCCC |
| C6H14 | |
| MFCD02179311 | |
| 8058 | |
| hexane |
Safety and Handling
ShelfLife : 1260 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Hexanes
EINECSNumber : 203-777-6
Recommended Storage : Room Temp