missing translation for 'onlineSavingsMsg'
Learn More

Hexane, CHROMASOLV™, for HPLC, ≥95%, Solstice

Catalog No. 6001042
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6001042 1 L Glass Bottle
6001044 2.5 L Glass Bottle
6001045 6 x 1 L Glass Bottle
60048241 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 6001042 Supplier Solstice Supplier No. 2705041L

CHROMASOLV™, for HPLC,≥95%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Melting Point -95°C (literature)
Absorbance max. 0.02 at 240 nm, max. 0.01 at 250 nm, max. 0.01 at 400 nm, max. 1.0 at 195 nm, max. 0.30 at 210 nm, max. 0.10 at 220 nm, max. 0.05 at 230 nm
CAS Min % ≥90.0000%
CAS Max % ≤100.0000%
Density 0.659 g/mL at 25 °C(lit.)
Quantity 1 L
Flash Point −26°C
Packaging Glass Bottle
Linear Formula CH3(CH2)4CH3
UN Number UN1208
Beilstein 1730733
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
Boiling Point 66°C to 69°C
Formula Weight 86.18
Percent Purity >95%
Grade HPLC
Chemical Name or Material Hexane, CHROMASOLV™, for HPLC, ≥95%
Show More Show Less
ShelfLife 1260 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Hexanes
EINECSNumber 203-777-6
Recommended Storage Room Temp